5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] Gemisch aus Isomeren fluoreszierender Farbstoff, Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: (5(6)-TAMRA SE

46406
PIER46406EA 182 EUR
PIER46406
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] Gemisch aus Isomeren fluoreszierender Farbstoff, Pierce™
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE]

NHS-rhodamine is a high performance derivative of rhodamine dye, activated for an amine-specific labelling of antibodies, proteins and other molecules.


  • Label antibodies for use as immunofluorescent probes
  • Label oligonucleotides for hybridization probes
  • Detect proteins in gels and on western blots
  • Excitation⁄Emission: 552⁄575 (nm)


NHS-rhodamine has a wide ranging application as antibody and other probe labels for use in fluorescence microscopy, flow cytometry, and immunofluorescence-based assays such as western blotting and ELISA.

Formel: C₂₉H₂₅N₃O₇
Molecular Weight: 527,52 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00077324
CAS-Nummer: 246256-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR