FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-isothiocyanatofluorescein, Fluorescein 5(6)-isothiocyanate, Fluorescein Isothiocyanate, Isomer I

46425
PIER46425EA 89.75 EUR
PIER46425
FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I)
FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I)
Formel: C₂₁H₁₁NO₅S
Molecular Weight: 389,381 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00005063
CAS-Nummer: 3326-32-7
EINECS: 222-042-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR