Pierce™ Spin Cups - Cellulose Acetate Filter
Lieferant: Thermo Fisher Scientific
69702
PIER69702EA 202 EUR
PIER69702
Pierce™ Spin Cups - Cellulose Acetate Filter
Filter Zentrifugaleinheiten
Pierce™ cellulose acetate filter spin cups are convenient tools for manipulating small volumes of affinity supports (20 - 400 µl) for protein purification. Simply add the affinity resin and sample to one of the spin cups, then use a microcentrifuge to efficiently wash away contaminants and elute the purified sample without losing any resin in the process. Spin cups allow more protein to be affinity purified in less time.
  • Cellulose acetate filters have smaller pore sizes than paper
  • Column volume: 800 µl
  • Resin volume: 20 to 400 µl
  • Filter type: Cellulose acetate filter, 0,45 µm pore size
  • Cap: Collection tube cap fits onto inserted spin cup
Applications include affinity purification/affinity chromatography, immunodepletion, immunoprecipitation (IP), and co-immunoprecipitation (co-IP).
Format: Microcentrifuge, spin column
Column type: Empty column