Pierce™ Spin Columns, Snap Cap
Lieferant: Thermo Fisher Scientific
69725
PIER69725EA 205 EUR
PIER69725
Pierce™ Spin Columns, Snap Cap
Filter Zentrifugaleinheiten
Pierce™ snap cap spin columns are convenient tools for manipulating small volumes of affinity supports (20 to 500 µl) for protein purification.
  • Column volume: 1 ml
  • Resin volume: 20 to 500 µl
  • Filter type: Polyethylene filter, ~30 µm pore size
  • Cap: Attached snap-cap (no cap on collection tube); press-on bottom caps
Applications include affinity purification/affinity chromatography, immunodepletion, immunoprecipitation (IP), and co-immunoprecipitation (co-IP).
Format: Microcentrifuge, spin column
Column type: Empty column