Freund's complete adjuvant, Imject™
Lieferant: Thermo Fisher Scientific
77140
PIER77140EA
147
EUR
PIER77140
Freund's complete adjuvant, Imject™
Protein- Expression
Proteinexpressions-Reagenzien
Imject™ Freund's Complete Adjuvant (FCA) is a popular non specific stimulator of the immune response for use in antibody production immunisation protocols.
- Greatly enhances immune response to immunogen
- Typically used for initial injections
- Water-in-oil emulsion containing killed Mycobacterium
Lieferumfang: Supplied ready to use in 10 ml ampoules.