Recombinant protein L, Pierce™

Lieferant: Thermo Fisher Scientific

21189 77679 77680
PIER21189EA 214 EUR
PIER21189 PIER77679 PIER77680
Recombinant protein L, Pierce™
Protein-Aufbereitungs Proteinreinigungs-Reagenzien

Purified (unconjugated) Pierce™ Recombinant Protein L is useful as the basis for preparing various kinds of probes or affinity media for detection or purification of mouse and human antibodies in immunoassays and antibody purification protocols.


  • Contains four immunoglobulin-binding domains per protein
  • Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
  • Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains


Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR