TCEP solution, neutral pH, Bond-Breaker™ Pierce™
Lieferant: Thermo Fisher Scientific
77720
PIER77720EA
197
EUR
PIER77720
TCEP solution, neutral pH, Bond-Breaker™ Pierce™
Protein-Aufbereitungs
Proteinreinigungs-Reagenzien
Bond-Breaker™ TCEP Solution (neutral pH) is a stable, 0,5 M solution of the thiol-free, phosphine-based TCEP compound, useful as a 10X stock for addition to SDS-PAGE sample loading buffers to reduce protein disulphide bonds.
- Odourless
- Selective and complete reduction of even the most stable water-soluble alkyl disulfides
- Reduces protein disulphides at room temperature and pH 5 in less than five minutes
- Resistant to air oxidation; nonvolatile and nonreactive toward other functional groups found in proteins