AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
78431
PIER78431EA
181
EUR
PIER78431
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
Formel:
C₈H₁₁ClFNO₂S Lagertemperatur: Kühlschrank |
MDL:
MFCD00132962 CAS-Nummer: 30827-99-7 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...