Subcellular protein fractionation kit for tissues
Lieferant: Thermo Fisher Scientific
87790 87791
87790.EA 658 EUR
87790. PIER87791
Subcellular protein fractionation kit for tissues
Puffer Extraktionspuffer
This kit is ideal for cell fractionation and enrichment of proteins from five different cellular compartments within two hours.
  • Optimized - formulations and protocols specific for fractionation of tissue
  • Complete - obtain cytoplasmic, membrane, soluble nuclear, chromatin-bound and cytoskeletal protein fractions from a single kit
  • Complete - obtain protein fractions from a single kit
  • Convenient - unique tissue strainer allows rapid removal of tissue debris
  • Versatile - extract proteins from a variety of tissue types, including liver, heart, brain, kidney, lung and spleen
  • Compatible - use extracts for downstream applications such as protein assays, Western blotting, protein assays, electrophoretic mobility shift assays, reporter-gene and enzyme-activity assays
Lieferumfang: Kit includes four extraction buffers, tissue strainers, a stabilized nuclease and halt protease inhibitor cocktail.