High capacity endotoxin removal spin columns, Pierce™
Lieferant: Thermo Fisher Scientific
88273 88274 88275 88276 88277
PIER88273EA 79.5 EUR
PIER88273 PIER88274 PIER88275 PIER88276 PIER88277
High capacity endotoxin removal spin columns, Pierce™
Filter Zentrifugaleinheiten
Pierce™ high capacity endotoxin removal spin columns are ideal for the removal of endotoxins from protein or antibody samples prior to cell transfection or animal injection.
  • Bind up to 2000000 EU/ml to eliminate grater than 99% of endotoxins
  • Recover greater than 85% of protein sample
  • Spin cup format enables endotoxin depletion within 1 hour
  • Single use spin columns avoid cross contamination of samples
This resin binds to reduce endotoxin levels in protein samples by ≥99% in one hour. Each column contains 25% slurry in 20% ethanol.