Spin filter plates, detergent removal, 96-well, Pierce™
Lieferant: Thermo Fisher Scientific
88304
PIER88304EA 782 EUR
PIER88304
Spin filter plates, detergent removal, 96-well, Pierce™
Mikrotiterplatten Mikrofiltrationsplatten
Pierce™ Detergent removal filter spin plates enable multiple-sample processing using a high-performance resin that offers effective detergent removal and excellent protein/peptide recovery. Pierce™ Detergent removal resin efficiently binds to and removes high concentrations of detergents from 10 µl to 1 ml samples with minimal sample loss.
  • Fast and efficient protocol
  • Removes greater than 95% of detergents with high protein recovery
  • Helps improves mass spectrometry peptide coverage
  • Best for processing samples with proteins or peptides at greater than 100 µg/ml
The Pierce™ Detergent removal resin specifically binds a wide variety of detergents and surfactants that are commonly used in protein extraction and biological sample preparation and provide an especially convenient and rapid format for treating protein and peptide solutions to remove interfering detergents before downstream analysis by mass spectrometry and other techniques.