Reserpine standard for LC-MS, Pierce™
Lieferant: Thermo Fisher Scientific
88326
PIER88326EA 188 EUR
PIER88326
Reserpine standard for LC-MS, Pierce™
Standards Farbreferenzen und Spektroskopie-Standards
Reserpine standard for LC-MS is a precise concentration of reserpine specifically designed for performance evaluation of mass spectrometers.
  • Pre-diluted and requires minimal additional dilutions to reach target concentration for injection
  • Easily removed and replaced with safe handling when withdrawing solution
  • Mass spectrometry grade reagent in a non leachable screw cap vials
The standard is provided in a concentration of 100 pg/µl in 50% isopropyl alcohol and requires minimal additional dilutions. The product is provided in a pack of 5×1 ml glass amber vials with PTFE-lined screw caps.