HeLa protein digest standard, Pierce™
Lieferant: Thermo Fisher Scientific
88328
88329
PIER88328EA
171
EUR
PIER88328
PIER88329
HeLa protein digest standard, Pierce™
Protein-Aufbereitungs
Proteinreinigungs-Reagenzien
The HeLa Protein Digest Standard is a highly validated mammalian protein digest that may be used as a quality control sample for mass spectrometry (MS) analysis of complex proteomic samples.
- Complex mammalian proteome sample protein digest (>15,000 proteins) from the HeLa S3 cell line
- Prepared using both LysC and trypsin to ensure less than 10% tryptic missed cleavages
- Validated peptide quality with <10% methionine oxidation and <10% lysine carbamylation
- High-quality, consistent protein digestion backed up by rigorous testing and documentation
- Provided in a stable lyophilised format