L(-)-Prolin SILAC
Lieferant: Thermo Fisher Scientific
Synonyme:
H-Pro-OH, (S)-(-)-Azolidin-2-carbonsäure, (S)-(-)-1,3,4,5-Tetrahydro-2H-pyrrol- 2-carbonsäure, (S)-(-)-Pyrrolidin-2-carbonsäure, (2S)-(-)-Pyrrolidin-2-carbonsäure, (S)-(-)-Prolin
88430
PIER88430EA
184
EUR
PIER88430
L(-)-Prolin SILAC
L(-)-Prolin
Formel:
C₅H₉NO₂ Molecular Weight: 115.13 g/mol Siedepunkt: 245 Schmelzpunkt: 220…222 °C Dichte: 1.35 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064318 CAS-Nummer: 147-85-3 EINECS: 205-702-2 Merck Index: 13,07871 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...