Ni-NTA magnetic beads, HisPur™, Pierce™
Lieferant: Thermo Fisher Scientific
88831 88832
PIER88831EA 246 EUR
PIER88831 PIER88832
Ni-NTA magnetic beads, HisPur™, Pierce™
Kugeln Magnetkugeln
The Pierce™ Ni-NTA magnetic beads are high-capacity nickel-IMAC beads for affinity purification of His-tagged fusion proteins in manual or automated formats.
  • Bead surface is pre-blocked and the protocol provides optimised buffers for purification
  • Fast, scalable and versitle to purify proteins using native or denaturing conditions
  • Can be use with common cell lysis reagents and a variety of buffer additives
  • Protein coupling to the beads and downstream applications can be performed manually or by automation