Isobaric mass tag labelling kit, iodoTMTsixplex™
Lieferant: Thermo Fisher Scientific
The iodoTMTsixplex™ Isobaric Mass Tag Labeling Kit contains iodoTMT™ reagents and associated materials for the irreversible labeling of sulfhydryl groups.
- Specifically reacts only with sulfhydryl groups (reduced, unmodified cysteine residues)
- Labeled proteins and peptides are not susceptible to reducing agents
- Options for duplex isotopic (MS) or sixplex isobaric (MS/MS) quantitation
- Can be used to study cysteine modifications (e.g. di-sulfide bridges, S-nitrosylation)
The iodoTMTsixplex™ Isobaric Mass Tag Labeling Kit enables concurrent identification and multiplexed quantitation of proteins in different samples using tandem mass spectrometry along with iodoTMTsixplex™ label reagents.
Lieferumfang: Kit includes 5×0,2 mg of iodoTMT6-126 label reagent, iodoTMT6-127 label reagent, iodoTMT6-128 label reagent, iodoTMT6-129 label reagent, iodoTMT6-130 label reagent, iodoTMT6-131 label reagent, HES buffer (15 ml), bond-breaker TCEP colution (0,5 ml), DTT No-Weigh™ format (8×7,7 mg), trypsin (5×20 µg), trypsin storage solution (250 µl) and bovine albumin (2,5 mg).