Hydroxylamin-Lösung 50% (w/w) in Wasser für TMT Experimente

Lieferant: Thermo Fisher Scientific
Gefahr

90115
PIER90115EA 89.5 EUR
PIER90115
Hydroxylamin-Lösung 50% (w/w) in Wasser für TMT Experimente
Hydroxylamin

Thermo Scientific 50% Hydroxylamine is a 10X quenching reagent for amine-reactive isobaric mass tag labeling experiments.


Used to prepare oximes, and is also an intermediate in biological nitrification.


Bitte beachten For research use only. Not for use in diagnostic procedures.

Formel: H₃NO
Molecular Weight: 33,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00044522
CAS-Nummer: 7803-49-8
EINECS: 232-259-2
UN: 3266
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

ASSAY/TITRATION 45.0 - 55.0% range
VISUAL Colorless/clear to slightly yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR