Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), No-Weigh™ Format, Pierce™
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Thermo Scientific Pierce Sulfo-SMCC, No-Weigh Format is a water-soluble, amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane spacer arm (8.3 angstroms).
- Reactive groups: sulfo-NHS ester and maleimide
- Reactive towards: amino and sulfhydryl groups
- Water-soluble (compare to SMCC), so crosslinking can be done in physiologic solutions
- High purity, crystalline reagent can be used to create high-purity maleimide-activated derivatives
- Cyclohexane bridge confers added stability to the maleimide group making Sulfo-SMCC the ideal crosslinking agent for maleimide activation of proteins. Maleimide groups are stable for 64 hours in 0.1 M sodium phosphate buffer, pH 7 at 4°C.
Sulfosuccinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate (Sulfo-SMCC) is a non-cleavable and membrane impermeable crosslinker. It contains an amine-reactive N-hydroxysuccinimide (NHS ester) and a sulfhydryl-reactive maleimide group. NHS esters react with primary amines at pH 7-9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. The maleimide groups of Sulfo-SMCC and SMCC are unusually stable up to pH 7.5 because of the cyclohexane bridge in the spacer arm. Because it contains the hydrophilic sulfonyl moiety, Sulfo-SMCC is soluble up to ∼10 mM in water and many commonly used buffers, thus avoiding the use of organic solvents which may perturb protein structure.
Bitte beachten For Research Use Only. Not for use in diagnostic procedures.
Formel:
C₁₆H₁₈N₂O₉S Schmelzpunkt: 179 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00054978 CAS-Nummer: 92921-24-9 |
Spezifikation Testergebnisse
Spacer Arm Length | Short, 8.3 angstroms |
Shipping Condition | Ambient |
Quantity | 10 x 2 mg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...