Natrium-1-hydroxy-2,5-dioxo-3-pyrrolidinsulfonat, No-Weigh™ Format, Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: N-Hydroxy sulphosuccinimide sodium salt, Hydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium salt, N-Hydroxysulphosuccinimide sodium salt, Sulfo NHS, Sulpho-N-Hydroxysuccinimide

A39269
PIERA39269EA 183 EUR
PIERA39269
Natrium-1-hydroxy-2,5-dioxo-3-pyrrolidinsulfonat, No-Weigh™ Format, Pierce™
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt)

Thermo Scientific Pierce Sulfo-NHS, No-Weigh Format is a chemical modification reagent for converting carboxyl groups to amine-reactive NHS esters for bioconjugation, crosslinking, labeling and immobilization methods.


  • Efficiency of EDC-mediated coupling is increased in the presence of Sulfo-NHS
  • Amine-reactive NHS esters or Sulfo-NHS esters can be made with any carboxyl-containing molecule
  • Sulfo-NHS derivatives are usually directly water-soluble (can be added directly to physiologic buffers) and membrane-impermable (can be used for cell surface labeling)
  • High-purity, crystalline Sulfo-NHS can be used to create high-quality activated derivatives
  • Purity—Greater than 95% by quantitative NMR (the highest standard for crosslinker purity); average lot purity is greater than 99%


Compared to the standard grade product, Premium Grade Sulfo-NHS provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-NHS (N-hydroxysulfosuccinimide) enables control and modification of carbodiimide crosslinking reactions involving activation of carboxylates (—COOH) for conjugation with primary amines (—NH2). Derivatives are easily synthesized by mixing the sulfo-NHS with a carboxyl-containing molecule and a dehydrating agent such as the carbodiimide EDC (EDAC). The method is the basis for generating many types of protein labeling reagents, including amine-reactive fluorescent dyes, biotin affinity tags and pegylation compounds.


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₄H₄NNaO₆S
Molecular Weight: 217.13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00043100
CAS-Nummer: 106627-54-7

Bestellen

Spezifikation Testergebnisse

Spacer Arm Length Short


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR