Cytology swabs, Histobrush®
Lieferant: PURITAN MEDICAL
25-2199
PMPC25-2199EA 2280 EUR
PMPC25-2199
Cytology swabs, Histobrush®
Tupfer Tupfer für klinische Verwendung
These cytology brushes feature tapered tips made out of soft nylon bristles and a long PS handle.
Used for the collection of cervical specimens during pap smear tests, as well as buccal cells, for DNA lineage.
Standards: ISO 9001:2008, 13485:2003, FDA GMP’s and 510K approved.
Bestellinformation: A certificate of compliance is available upon request.