Tubing connectors
Lieferant: CADENCE SCIENCE
6169IND
POPR6169INDEA 34.3 EUR
POPR6169IND
Tubing connectors
Fittings und Verbinder
These tubing connectors are used in hospitals, laboratories, and biomedical instruments. The metal connectors are precision machined and are made from either nickel plated brass or 316 stainless steel.
Standard luer tapers meets ISO 594 specifications include Female Luer Lock (FLL), Micro-Mate® Female Luer Lock (MM FLL), Male Luer Lock (MLL) and Male Luer (ML).