Mop frame, stainless steel, Clino® CR
Lieferant: PFENNIG REINIGUNGSTECHNIK
2009061
PPSA2009061EA 106 EUR
PPSA2009061
Mop frame, stainless steel, Clino® CR
Mopps
Robust and durable through stainless steel mop frame.
  • Autoclavable
  • Contactless release of the mop with MopDrop
  • With quick-change system ClinoConnect