Cleaning trolley Clino®, E1e
Lieferant: PFENNIG REINIGUNGSTECHNIK
2020400
PPSA2020400EA 1220 EUR
PPSA2020400
Cleaning trolley Clino®, E1e
Transportwagen
The Clino® E1e is the smallest possible cleaning trolley system with stainless steel flat wringer with electropolished base module suitable for high sensitive areas that require minimal space.
  • Ergonomic design
  • GMP conformity
  • Full flexibility in single- and two-step technique
  • Has double ball-bearinged guide wheels 75 mm
Lieferumfang: Supplied with 2×plastic blue buckets (25 and 15 L).