Cleanroom mops, King
Lieferant: PFENNIG REINIGUNGSTECHNIK
2140020
PPSA2140020EA 16.9 EUR
PPSA2140020
Cleanroom mops, King
Mopps
Mops with high area performance.
  • Provides maximum surface capacity and excellent dirt pick-up capability
  • Reference in the single or dual-stage process
  • Ready-to-use pre-washed
  • Made up of synthetic/viscose yarn (50:50)
  • With cover sheet, PVC pocket and sewed up double-latching pocket