Cleanroom mops, MicroKing
Lieferant: PFENNIG REINIGUNGSTECHNIK
2140080
PPSA2140080EA 20.5 EUR
PPSA2140080
Cleanroom mops, MicroKing
Mopps
Mops for grey areas and for all water-resistant hard floor coverings.
  • Increased cleaning performance and maximum dirt absorption facilities
  • Highest area performance and best tuft-quality
  • Made up of synthetic/viscose yarn (50:50) with additional microfiber lines
  • With cover sheet and PVC pocket, sewed up double-latching pocket
  • Ready-to-use, pre-washed
For the use in ISO class 9.