Cleanroom mops, SilberKing
Lieferant: PFENNIG REINIGUNGSTECHNIK
2160000
PPSA2160000EA 24.4 EUR
PPSA2160000
Cleanroom mops, SilberKing
Mopps
Mops for all hygienically sensitive areas, buildings without dryer.
  • Suitable in areas in which mops need to be stored for longer times
  • For impregnation of mop covers without disinfectants
  • Ready-to-use pre-washed
  • Made up of synthetic/viscose yarn (50:50)
  • With cover sheet, silver, PVC pocket and sewed up double-latching pocket