Cleanroom mops, MicroMopp Plus
Lieferant: PFENNIG REINIGUNGSTECHNIK
2700530
PPSA2700530EA 0 EUR
PPSA2700530
Cleanroom mops, MicroMopp Plus
Mopps
Mops for wet and moist cleaning in single-stage technique.
  • For all smooth and hard floor coverings
  • Made up of high-tech-microfiber 100% PES
  • With backfilling, cover sheet, loop with TEC-polyester and pocket of harness for extremely tear-proof
For the use in ISO class 9.