Cleanroom mops, classic mop
Lieferant: PFENNIG REINIGUNGSTECHNIK
2750070
PPSA2750070EA 9.8 EUR
PPSA2750070
Cleanroom mops, classic mop
Mopps
The classic cotton mop with textile pocket.
  • Finely prewashed mops
  • Made up of cotton/poiyester (35:65)