Squeegee mop and handles
Lieferant: PFENNIG REINIGUNGSTECHNIK
3590210 3400715 3590200
PPSA3590210EA 134 EUR
PPSA3590210 PPSA3400715 PPSA3590200
Squeegee mop and handles
Mopps
Squeegee mops and handles for hygienically sensitive areas.
  • Mop: PP (body), TPE (lip)
  • Hygienic squeegee is suitable with cleanroom and telescopical handles
  • Handles are anodised aluminum tubes