Cleanroom mops, standard, MicroMopp CR/A
Lieferant: PFENNIG REINIGUNGSTECHNIK
3500067
PPSA3500067EA 26.1 EUR
PPSA3500067
Cleanroom mops, standard, MicroMopp CR/A
Mopps
MicroMopp Standard CR/A mops are special multiple-use cleanroom mops.
  • Completely closed processing of polyester microsnares
  • 100 % PES-microfiber with polypropylene bristle
  • Rectangular mop design to reach edges perfectly well
  • Very good slide characteristics, not suitable for chlorinated disinfectants
  • For dissolving adhering contaminations and suitable for impregnation with flat wringer
  • With water-permeable cover and PU backfilling four enormous sqm-performance
  • Polyester pocket with drainage function