Clino® OneWay Disposable Mop
Lieferant: PFENNIG REINIGUNGSTECHNIK
3500947
PPSA3500947EA 25.7 EUR
PPSA3500947
Clino® OneWay Disposable Mop
Mopps
One way disposable mop for cleanroom sword CR/A.
  • Cleaning of hard to reach areas
  • Autoclavable, disinfectant resistant
Working material: 70% polyester (microfibre), 30% polyester (white), loop 100% polyprobylene