Cleanroom wipes, Clino® one way
Lieferant: PFENNIG REINIGUNGSTECHNIK
3600070
PPSA3600070EA 72.8 EUR
PPSA3600070
Cleanroom wipes, Clino® one way
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
Clino® one way clean surfaces in semiconductors and pharma industries.
  • More excellent intake capacity
  • High abrasion resistance
  • Low particle and fibre emission
Suitable for use in ISO class 7 and 8 (GMP grade C and D) cleanrooms.