Temperature and humidity loggers, Hygro Button
Lieferant: PROGES PLUS
DAL0084
PRGPDAL0084EA 287 EUR
PRGPDAL0084
Temperature and humidity loggers, Hygro Button
Datenlogger
Small and rugged, the Hygro Button data loggers can survive any harsh environment. They are suitable for temperature and humidity control, product tracking, transport, process analysis, HACCP. Ideal in controlling temperature and humidity in cellars, dairy industry and also in stability tests, chambers and museums.
  • Collect and record date and time
  • Programmable low and high alarm threshold with temporization parameters
Ø×H: 16×6 mm
Sample rate: 1 sec - 273 hr
Standards: Conform to IP 65.