Ammoniaklösung 25% 100 ml/l solution

Lieferant: PROLABO CMP
Gefahr

Synonyme: Ammoniumhydroxid, Ammoniak

85163.410
PRLS85163.410EA 0 EUR
PRLS85163.410
Ammoniaklösung 25% 100 ml/l solution
Ammoniaklösung

Concentrated solution to dilute to 100 mL/L.

Formel: NH₃
Molecular Weight: 35,05 g/mol
Dichte: 0,92 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066650
CAS-Nummer: 1336-21-6
EINECS: 215-647-6
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR