Benzo[k]fluoranthen

Lieferant: LGC Standards PROMOCHEM

BCR-048R
PROOBCR-048REA 165 EUR
PROOBCR-048R
Benzo[k]fluoranthen
Benzo[k]fluoranthen
Formel: C₂₀H₁₂
MDL: MFCD00046287
CAS-Nummer: 207-08-9
EINECS: 205-916-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR