1-Methylbenz[a]anthracen

Lieferant: LGC Standards PROMOCHEM

BCR-093R
PROOBCR-093REA 162 EUR
PROOBCR-093R
1-Methylbenz[a]anthracen
1-Methylbenz[a]anthracen
Formel: C₁₉H₁₄
CAS-Nummer: 2498-77-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR