Benzo[a]fluoranthen

Lieferant: LGC Standards PROMOCHEM

BCR-097
PROOBCR-097EA 564 EUR
PROOBCR-097
Benzo[a]fluoranthen
Benzo[a]fluoranthen
Formel: C₂₀H₁₂
CAS-Nummer: 203-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR