Dibenzo[a,i]acridin

Lieferant: LGC Standards PROMOCHEM

BCR-152
PROOBCR-152EA 327 EUR
PROOBCR-152
Dibenzo[a,i]acridin
Dibenzo[a,i]acridin
Formel: C₂₁H₁₃N
Molecular Weight: 279.34 g/mol
CAS-Nummer: 226-92-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR