Dibenzo[a,h]acridin

Lieferant: LGC Standards PROMOCHEM

BCR-153R
PROOBCR-153REA 162 EUR
PROOBCR-153R
Dibenzo[a,h]acridin
Dibenzo[a,h]acridin
Formel: C₂₁H₁₃N
Molecular Weight: 279.34 g/mol
CAS-Nummer: 226-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR