Dibenzo[a,c]acridin
Lieferant: LGC Standards PROMOCHEM
BCR-155
PROOBCR-155EA
573
EUR
PROOBCR-155
Dibenzo[a,c]acridin
Dibenzo[a,c]acridin
Formel:
C₂₁H₁₃N Molecular Weight: 279.34 g/mol |
CAS-Nummer:
215-62-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...