Dibenzo[c,h]acridin

Lieferant: LGC Standards PROMOCHEM

BCR-156R
PROOBCR-156REA 161 EUR
PROOBCR-156R
Dibenzo[c,h]acridin
Dibenzo[c,h]acridin
Formel: C₂₁H₁₃N
Molecular Weight: 279.34 g/mol
CAS-Nummer: 224-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR