Benzo[a]acridin

Lieferant: LGC Standards PROMOCHEM

BCR-157
PROOBCR-157EA 573 EUR
PROOBCR-157
Benzo[a]acridin
Benzo[a]acridin
Formel: C₁₇H₁₁N
CAS-Nummer: 225-11-6
EINECS: 205-929-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR