Benzo[c]acridin

Lieferant: LGC Standards PROMOCHEM

BCR-158
PROOBCR-158EA 573 EUR
PROOBCR-158
Benzo[c]acridin
Benzo[c]acridin
Formel: C₁₇H₁₁N
CAS-Nummer: 225-51-4
EINECS: 205-930-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR