Certified reference material, soya-maize oil blend, BCR®
Lieferant: LGC Standards PROMOCHEM
BCR-162R
PROOBCR-162REA
260
EUR
PROOBCR-162R
Certified reference material, soya-maize oil blend, BCR®
Standards
Lebensmittel-Standards
A unit contains approximately 5,5 gm soya-maize oil blend in 10 ml amber ampoule.
- Fatty acid analysis
Standards: IRMM certified.
Bitte beachten Store in Freezer below –20 °C.