Aluminiumoxid, α-Phase

Lieferant: LGC Standards PROMOCHEM

BCR-169
PROOBCR-169EA 211 EUR
PROOBCR-169
Aluminiumoxid, α-Phase
Aluminiumoxid, α-Phase
Formel: Al₂O₃
MDL: MFCD00003424
CAS-Nummer: 1344-28-1
EINECS: 215-691-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR