2,4,4'-Trichlorbiphenyl (PCB Nr. 28)

Lieferant: LGC Standards PROMOCHEM

Synonyme: 2,4,4'-PCB, PCB Nr. 28, 1-(4-Chlorphenyl)-2,4-dichlorbenzol

BCR-291
PROOBCR-291EA 414 EUR
PROOBCR-291
2,4,4'-Trichlorbiphenyl (PCB Nr. 28)
2,4,4'-Trichlorbiphenyl (PCB Nr. 28)
Formel: C₁₂H₇Cl₃
Molecular Weight: 257.55 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 7012-37-5
EINECS: 230-293-2
UN: 2315
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR