2,2',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 153)
Lieferant: LGC Standards PROMOCHEM
BCR-297
PROOBCR-297EA
410
EUR
PROOBCR-297
2,2',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 153)
2,2',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 153)
Formel:
C₁₂H₄Cl₆ Molecular Weight: 360.88 g/mol Siedepunkt: 259 Schmelzpunkt: 103…104 °C Lagertemperatur: Kühlschrank |
CAS-Nummer:
35065-27-1 UN: 3432 ADR: 9 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...