Standards for formaldehyde-2,4-dinitrophenyl-hydrazone, BCR®
Lieferant: LGC Standards PROMOCHEM
BCR-553-4
PROOBCR-553-4EA 0 EUR
PROOBCR-553-4
Standards for formaldehyde-2,4-dinitrophenyl-hydrazone, BCR®
Standards Umweltanalyse-Standards
Used on glass fibre filters.
Bestellinformation: The kit consists of 2 samples of BCR-553 (spiked filter) and 1 sample of BCR-554 (blank filter).