Acepromazinmaleat

Lieferant: LGC Standards PROMOCHEM

BP001
PROOBP001EA 255 EUR
PROOBP001
Acepromazinmaleat
Acepromazinmaleat
Formel: C₂₃H₂₆N₂O₅S
Molecular Weight: 442,53 g/mol
CAS-Nummer: 3598-37-6
EINECS: 222-748-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR