Ethosuximid

Lieferant: LGC Standards PROMOCHEM

Synonyme: 2-Ethyl-2-methylsuccinimide, 3-Ethyl-3-methylpyrrolidine-2,5-dione

BP146
PROOBP146EA 297 EUR
PROOBP146
Ethosuximid
Ethosuximid
Formel: C₇H₁₁NO₂
Molecular Weight: 141,17 g/mol
MDL: MFCD00072123
CAS-Nummer: 77-67-8
EINECS: 201-048-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR